Anti-Integrin alpha 4 Rabbit Polyclonal Antibody
Catalog # ANTIA95689-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:269C wild type
- Antigen symbol:Integrin alpha 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P13612
- Antigen synonyms:CD49d|Antigen CD49D, alpha 4 subunit of VLA 4 receptor|CD49 antigen-like family member D|CD49 antigen like family member D|Integrin alpha IV|IA4|Integrin alpha-4|Integrin alpha 4 subunit|Integrin alpha 4
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:115 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human Integrin alpha4 (amino acids 983-1032)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to Integrin alpha4 for WB and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Integrin alpha 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse