Order Entry
Netherlands
ContactUsLinkComponent
Anti-Enterovirus 71 3D Rabbit Monoclonal Antibody [clone: ARC58400]
Anti-Enterovirus 71 3D Rabbit Monoclonal Antibody [clone: ARC58400]
Catalog # ANTIA309755-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Enterovirus 71 3D Rabbit Monoclonal Antibody [clone: ARC58400]
Catalog # ANTIA309755-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Enterovirus 71 3D
  • Antigen symbol:
    Enterovirus 71 3D
  • Clonality:
    Monoclonal
  • Clone:
    ARC58400
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of Enterovirus EV71 3D.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC58400] antibody to Enterovirus 71 3D for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:2,000 to 1:20,000

Type: Primary
Antigen: Enterovirus 71 3D
Clonality: Monoclonal
Clone: ARC58400
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human