Anti-ALK Rabbit Monoclonal Antibody [clone: ARC58012]
Catalogus # ANTIA309279-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ALK
- Antigen symbol:ALK
- Clonality:Monoclonal
- Clone:ARC58012
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9UM73
- Antigen synonyms:ALK/EML4 fusion gene, included|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK tyrosine kinase receptor|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
- Molecular weight:140 kDa/220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human ALK.
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC58012] antibody to ALK for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: ARC58012
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat