Anti-Synaptotagmin Rabbit Polyclonal Antibody
Catalog # ANTIA309002-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HsT1192
- Antigen symbol:Synaptotagmin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9BQG1
- Antigen synonyms:SVP65|Synaptotagmin II|P65|Synaptotagmin V|Synaptotagmin IV|synaptotagmin 4|Synaptotagmin|Synaptotagmin I|MYSPC
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% thiomersal
- Molecular weight:63 kDa
- Sequence:TVTIIKASNLKAMDLTGFSDPYVKASLISEGRRLKKRKTSIKKNTLNPTYNEALVFDVAPESVENVGLSIAVVDYDCIGHNEVIGVCRVGPDAADPHGREHWAEMLANPRKPVEHWHQLVEEKTVTSFTKGSKGLSEKENSE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 449-590 of human SYT3 (NP_115674.1).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Synaptotagmin for WB with samples derived from mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Synaptotagmin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat