Anti-Semaphorin 3F Rabbit Polyclonal Antibody
ANTIA96744-100
New Product
- Antibody type:Primary
- Antigen name:SEM3F_HUMAN
- Antigen symbol:Semaphorin 3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q13275
- Antigen synonyms:Sema IV|sema domain immunoglobulin domain Ig short basic domain secreted semaphorin 3F|sema domain immunoglobulin domain Ig short basic domain secreted 3F|SemaIII/F|SemaIV|SEMA-IV|SEMA4|Sema III/F|SEMA3F
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:88 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human SEMA3F (amino acids 734-783)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to SEMA3F for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Semaphorin 3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat