Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-40]
Catalog # ANTIA85917-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HLA DP
- Antigen symbol:HLA DR + HLA DP
- Clonality:Monoclonal
- Clone:HL-40
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Pig,Human
- Form:Liquid
- Antigen synonyms:HLA DR + HLA DP|MHC Class II|HLA DR
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji Burkitt's lymphoma cell line
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HL-40] antibody to HLA DR + HLA DP for flow cytometry, IP and IHC-P with samples derived from human and porcine.
- Validated applications: Flow cytometry, IP, IHC-P
- Recommended dilutions: Flow cytometry: 1 µg/ml
Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: HL-40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine