Order Entry
Netherlands
ContactUsLinkComponent
Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA85911-100
Supplier:  Antibodies.com
New Product
Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA85911-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BDPLT16
  • Antigen symbol:
    Integrin beta 3
  • Clonality:
    Monoclonal
  • Clone:
    VIPL2
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05106
  • Antigen synonyms:
    HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Unfortunately, this information is considered to be commercially sensitive
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [VIPL2] antibody to Integrin beta 3 (FITC) for flow cytometry with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: VIPL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates