Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: MEM-136] (APC (Allophycocyanin))
Catalog # ANTIA85626-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HLA DP
- Antigen symbol:HLA DR + HLA DP
- Clonality:Monoclonal
- Clone:MEM-136
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:HLA DR + HLA DP|MHC Class II|HLA DR
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:PHA-activated peripheral blood lymphocytes
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-136] antibody to HLA DR + HLA DP (APC) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: MEM-136
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human