Anti-CD99 Mouse Monoclonal Antibody [Clone: 3B2/TA8] (FITC (Fluorescein Isothiocyanate))
ANTIA85605-100
New Product
- Antibody type:Primary
- Antigen name:12E7
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:3B2/TA8
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|CD99 molecule|Cell surface antigen O13|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [3B2/TA8] antibody to CD99 (FITC) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 3B2/TA8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human