Order Entry
Netherlands
ContactUsLinkComponent
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: W6/32] (Biotin)
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: W6/32] (Biotin)
Catalog # ANTIA85489-100
Supplier:  Antibodies.com
New Product
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: W6/32] (Biotin)
Catalog # ANTIA85489-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp686P19218
  • Antigen symbol:
    HLA Class I
  • Clonality:
    Monoclonal
  • Clone:
    W6/32
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Non-human primates,
    Human,
    Bovine,
    Cat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Store at 2...8 °C. Do not freeze.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Membrane of human tonsil cells
  • Tested applications:
    IHC-Fr, ICC
  • Purification:
    This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [W6/32] antibody to HLA Class I (Biotin) for flow cytometry, IP, WB, IHC-Fr, ICC and ELISA with samples derived from human, bovine, feline and non-human primates.

  • Validated applications: Flow cytometry, IP, WB, IHC-Fr, ICC, ELISA
  • Recommended dilutions: Flow cytometry: 1 to 12 µg/ml

Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Bovine, Cat, Non-Human Primates