Order Entry
Netherlands
ContactUsLinkComponent
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147]
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147]
Catalog # ANTIA85472-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147]
Catalog # ANTIA85472-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp686P19218
  • Antigen symbol:
    HLA Class I
  • Clonality:
    Monoclonal
  • Clone:
    MEM-147
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    PHA-activated peripheral blood lymphocytes
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-147] antibody to HLA Class I for flow cytometry, IP and WB with samples derived from human.

  • Validated applications: Flow cytometry, IP, WB
  • Recommended dilutions: Flow cytometry: 1 µg/ml

Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: MEM-147
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human