Anti-USP43 Goat Polyclonal Antibody
ANTIA84543-100
New Product
- Antibody type:Primary
- Antigen name:Deubiquitinating enzyme 43
- Antigen symbol:USP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Ubiquitin specific protease 43|Ubiquitin thiolesterase 43|Ubiquitin-specific-processing protease 43|USP43|UBP43|Ubiquitin carboxyl-terminal hydrolase 43|Ubiquitin thioesterase 43|UBP43_HUMAN|ubiquitin specific peptidase 43
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-SQRTVPGEQASYGT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human USP43 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to USP43 for ELISA and IHC with samples derived from human.
- Validated applications: ELISA, IH
Type: Primary
Antigen: USP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human