Order Entry
Netherlands
ContactUsLinkComponent
Anti-BDH2 Goat Polyclonal Antibody
Anti-BDH2 Goat Polyclonal Antibody
Catalog # ANTIA84456-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-BDH2 Goat Polyclonal Antibody
Catalog # ANTIA84456-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    3 hydroxybutyrate dehydrogenase type 2
  • Antigen symbol:
    BDH2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    Dehydrogenase/reductase SDR family member 6|BDH2_HUMAN|EFA6R|BDH2|Dehydrogenase/reductase (SDR family) member 6|DHRS6|BDH 2|3-hydroxybutyrate dehydrogenase type 2|FLJ13261
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-TKKKQIDQFANE
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human BDH2 (internal region)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to BDH2 for ELISA, WB and IHC with samples derived from human.

  • Validated applications: ELISA, WB, IH

Type: Primary
Antigen: BDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human