Anti-Junctional Adhesion Molecule 1 Rabbit Polyclonal Antibody
ANTIA99902-100
New Product
- Antibody type:Primary
- Antigen name:CD 321
- Antigen symbol:Junctional Adhesion Molecule 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9Y624
- Antigen synonyms:ESTM33|CD321|JAM-1|F11 receptor|JAM A|F11R|JAM|CD321 antigen|JAM 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:32 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human F11R (amino acids 191-240)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to F11R for WB and ELISA with samples derived from human and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Junctional Adhesion Molecule 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat