Anti-USP16 Goat Polyclonal Antibody
Catalogus # ANTIA83203-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Deubiquitinating enzyme 16
- Antigen symbol:USP16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Ubiquitin thiolesterase 16|MSTP039|Ubiquitin carboxyl-terminal hydrolase 16|Ubiquitin Specific Protease 16|Ubiquitin-specific-processing protease 16|Ubiquitin Specific peptidase 16|UBP M|Human ubiquitin processing protease|Ubiquitin-processing protease UBP-M
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:GKKRTKGKTVPIDD-C
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human USP16 (N terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to USP16 for ELISA and IHC with samples derived from human.
- Validated applications: ELISA, IHC
Type: Primary
Antigen: USP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human