Anti-ALDH18A1 Goat Polyclonal Antibody
ANTIA82892-100
New Product
- Antibody type:Primary
- Antigen name:2810433K04Rik
- Antigen symbol:ALDH18A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:AI429789|Delta1 pyrroline 5 carboxlate synthetase|Aldehyde dehydrogenase 18 family member A1|Aldehyde dehydrogenase family 18 member A1|ALDH18A1|Gamma-glutamyl kinase|Gamma-glutamyl phosphate reductase|Delta 1 pyrroline 5 carboxylate synthetase|Aldehyde dehydrogenase 18A1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-SEHGSLKYLH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ALDH18A1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to ALDH18A1 for ELISA and WB with samples derived from human and mouse.
- Validated applications: ELISA, WB
Type: Primary
Antigen: ALDH18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse