Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-NME2 Mouse Monoclonal Antibody [clone: CPTC-NME2-2]
  ANTIA252704-100
New Product
undefined
Anti-NME2 Mouse Monoclonal Antibody [clone: CPTC-NME2-2]
  ANTIA252704-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    C myc purine binding transcription factor PUF
  • Antigen symbol:
    NME2
  • Clonality:
    Monoclonal
  • Clone:
    CPTC-NME2-2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P22392
  • Antigen synonyms:
    epididymis secretory sperm binding protein Li 155an|C-myc purine-binding transcription factor PUF|HEL-S-155an|Histidine protein kinase NDKB|C myc transcription factor|NDKB_HUMAN|NDK B|MGC111212|MGC2212
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    17 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human NME2 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [CPTC-NME2-2] antibody to NME2 for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: CPTC-NME2-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human