Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/1495R]
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/1495R]
Catalog # ANTIA252514-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD99 Rabbit Recombinant Antibody [clone: MIC2/1495R]
Catalog # ANTIA252514-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell surface antigen O13
  • Antigen symbol:
    CD99
  • Clonality:
    Recombinant
  • Clone:
    MIC2/1495R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P14209
  • Antigen synonyms:
    CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|120000000|CD99 molecule|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    27 - 32 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human MIC2 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MIC2/1495R] antibody to CD99 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 5 to 10 µg/ml

Type: Primary
Antigen: CD99
Clonality: Recombinant
Clone: MIC2/1495R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human