Anti-Ubiquitin Rabbit Recombinant Antibody [clone: UBB/3143R]
Catalog # ANTIA250263-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Epididymis secretory protein Li 50
- Antigen symbol:Ubiquitin
- Clonality:Recombinant
- Clone:UBB/3143R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P0G47, P0G48, P62979, P62987, P62988
- Antigen synonyms:UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:9 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1 - 119, of human Ubiquitin protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [UBB/3143R] antibody to Ubiquitin for IHC-P with samples derived from Human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Ubiquitin
Clonality: Recombinant
Clone: UBB/3143R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human