Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-TRIM27 Mouse Monoclonal Antibody [clone: PCRP-TRIM27-1B3]
  ANTIA253064-100
New Product
undefined
Anti-TRIM27 Mouse Monoclonal Antibody [clone: PCRP-TRIM27-1B3]
  ANTIA253064-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AW538890
  • Antigen symbol:
    TRIM27
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-TRIM27-1B3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P14373
  • Antigen synonyms:
    MGC189472|DAAP-182E11.3|Rfp1|RNF76|RING finger protein 76|RFP transforming protein|OTTHUMP00000029314|Ret finger protein|RFP
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    58,49 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human TRIM27 protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [PCRP-TRIM27-1B3] antibody to TRIM27 for Flow Cytometry and IF with samples derived from Human.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: TRIM27
Clonality: Monoclonal
Clone: PCRP-TRIM27-1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human