Order Entry
Netherlands
ContactUsLinkComponent
Anti-Keratin 76 Rabbit Recombinant Antibody [clone: KRTH/1576R]
Anti-Keratin 76 Rabbit Recombinant Antibody [clone: KRTH/1576R]
Catalog # ANTIA251011-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Keratin 76 Rabbit Recombinant Antibody [clone: KRTH/1576R]
Catalog # ANTIA251011-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Keratin 76
  • Clonality:
    Recombinant
  • Clone:
    KRTH/1576R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q01546
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    52 - 67 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human KRT76 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRTH/1576R] antibody to Keratin 76 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 0,25 to 0,5 µg/ml

Type: Primary
Antigen: Keratin 76
Clonality: Recombinant
Clone: KRTH/1576R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human