Anti-CD47 Rabbit Recombinant Antibody [clone: CD47/6362R]
Catalog # ANTIA250733-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:CD47
- Clonality:Recombinant
- Clone:CD47/6362R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q08722
- Antigen synonyms:IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:~50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 18 - 135, of human CD47 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CD47/6362R] antibody to CD47 for ELISA, WB and IHC-P with samples derived from Human.
- Validated applications: ELISA, WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD47
Clonality: Recombinant
Clone: CD47/6362R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human