Anti-ALK Rabbit Polyclonal Antibody
ANTIA10983-100
New Product
- Type antilichaam:Primair
- antigeen symbool:ALK
- Clonality:Polyklonaal
- Conjugatie:Onconjugeerd
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Formulier:Liquid
- Gen-ID:UniprotID# Q9UM73
- Antigeen synoniemen:ALK/EML4 fusion gene, included|ALK|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
- Modification:phospho Tyr1096
- Opslagbuffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Moleculair gewicht:168 kDa
- Sequentie:PNYCF
- Opslagtemperatuur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogeen:A synthetic phosphorylated peptide around Y1096 of human ALK (NP_004295.2)
- Purification:Affinity purification
- Verpakking:Plastic vial
- VPE:100 µl
Rabbit polyclonal antibody to ALK (phospho Tyr1096) for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: ALK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human