Anti-CD10 Mouse Monoclonal Antibody [clone: MEM-78] (PE-Cyanine 5)
ANTIA122040-100
New Product
- Antibody type:Primary
- Antigen name:Atriopeptidase
- Antigen symbol:CD10
- Clonality:Monoclonal
- Clone:MEM-78
- Conjugation:PE (Phycoerythrin)-Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08473
- Antigen synonyms:Enkephalinase|CD10|Common acute lymphocytic leukemia antigen|DKFZp686O16152|CALLA|Membrane metallo endopeptidase|EC 3.4.24.11|CD10 antigen|EPN
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:NALM-6 human pre-B cell line
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 5 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [MEM-78] antibody to CD10 (PE-Cyanine 5) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: MEM-78
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human