Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 7)
Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 7)
Catalog # ANTIA122014-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: MEM-28] (PE-Cyanine 7)
Catalog # ANTIA122014-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B220
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    MEM-28
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08575
  • Antigen synonyms:
    CD45R|CD45|GP180|L-CA|Leukocyte Common antigen|LCA|loc|CD45 antigen|CD 45
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes and T lymphocytes
  • Purification:
    This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-28] antibody to CD45 (PE-Cyanine 7) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human