Anti-CD21 Mouse Monoclonal Antibody [clone: LT21] (PerCP (Peridinin-Chlorophyll Protein Complex))
ANTIA121878-100
New Product
- Antibody type:Primary
- Antigen name:C3DR
- Antigen symbol:CD21
- Clonality:Monoclonal
- Clone:LT21
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine,Dog
- Form:Liquid
- Gene ID:UniprotID# P20023
- Antigen synonyms:Complement C3d receptor|CR|Complement component (3d/Epstein Barr virus) receptor 2|CD 21|Complement C3d receptor 2|Complement receptor type 2|CD21|Cr2|CR2_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:IM9 human B-lymphoblastoid cell line
- Purification:This antibody is conjugated with PerCP under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [LT21] antibody to CD21 (PerCP) for Flow Cytometry with samples derived from Porcine, Human, Canine and Bovine.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: LT21
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Porcine;Human;Dog;Bovine