Order Entry
Netherlands
ContactUsLinkComponent
Anti-GRIN2A Goat Polyclonal Antibody
Anti-GRIN2A Goat Polyclonal Antibody
Catalog # ANTIA121167-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-GRIN2A Goat Polyclonal Antibody
Catalog # ANTIA121167-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EPND
  • Antigen symbol:
    GRIN2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Antigen synonyms:
    hNR2A|LKS|GRIN2A|GluN2A|GRIN 2A|Glutamate [NMDA] receptor subunit epsilon-1|FESD|Glutamate receptor|Glutamate receptor ionotropic N methyl D aspartate 2A
  • Storage buffer:
    Supplied in Tris Buffered Saline, pH 7,30, with 0,02% Sodium Azide and 0,5% BSA
  • Sequence:
    C-EDAKTQVQLKKIHS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human GRIN2A (internal region)
  • Purification:
    Purified from goat serum by a mMonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to GRIN2A for ELISA and IF with samples derived from mouse.

  • Validated applications: ELISA, IF

Type: Primary
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse