Anti-PGP9.5 Rabbit Recombinant Antibody [clone: UCHL1/7076R]
Catalog # ANTIA316898-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Epididymis luminal protein 117
- Antigen symbol:PGP9.5
- Clonality:Recombinant
- Clone:UCHL1/7076R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P09936
- Antigen synonyms:HEL 117|Neuron cytoplasmic protein 9.5|HEL S 53|Epididymis secretory protein Li 53|OTTHUMP00000218141|OTTHUMP00000218139|OTTHUMP00000218140|NDGOA|OTTHUMP00000218137
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:20 - 30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human UCHL1 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [UCHL1/7076R] antibody to PGP9.5 for WB and IHC-P with samples derived from human and Rat.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 2 to 4 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: PGP9.5
Clonality: Recombinant
Clone: UCHL1/7076R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human;Rat