Anti-Myeloperoxidase Rabbit Recombinant Antibody [clone: MPO/8997R]
Catalog # ANTIA316739-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:84 kDa myeloperoxidase
- Antigen symbol:Myeloperoxidase
- Clonality:Recombinant
- Clone:MPO/8997R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05164
- Antigen synonyms:fj80f04|myeloid-specific peroxidase|mpx|89 kDa myeloperoxidase|Myeloperoxidase light chain|Myeloperoxidase heavy chain|MPO|EC 1.11.1.7|EC1.11.2.2
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:heavy - light promoter: 72 kDa/dimer: 140 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human MPO protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MPO/8997R] antibody to Myeloperoxidase for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Myeloperoxidase
Clonality: Recombinant
Clone: MPO/8997R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human