Order Entry
Netherlands
ContactUsLinkComponent
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Catalog # ANTIA316597-100
Leverancier:  Antibodies.com
New Product
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Catalog # ANTIA316597-100
Leverancier:  Antibodies.com
New Product

Specificaties

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 77
  • Clonality:
    Recombinant
  • Clone:
    rKRTL/8751
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Turtle,
    Rabbit,
    Monkey,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q7Z794
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    40 - 56,5 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Human epidermal keratin.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specificaties

Over dit artikel

Recombinant mouse monoclonal [rKRTL/8751] antibody to Keratin 77 for IHC-P with samples derived from human, monkey, bovine, canine, rabbit, mouse, rat, chicken and turtle.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 0,5 to 1 µg/ ml

Type: Primary
Antigen: Keratin 77
Clonality: Recombinant
Clone: rKRTL/8751
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Bovine;Dog;Rabbit;Mouse;Rat;Chicken;Turtle