Order Entry
Netherlands
ContactUsLinkComponent
Anti-Keratin 77 Rabbit Recombinant Antibody [clone: KRTL/7784R]
Anti-Keratin 77 Rabbit Recombinant Antibody [clone: KRTL/7784R]
Catalog # ANTIA316596-100
Supplier:  Antibodies.com
New Product
Anti-Keratin 77 Rabbit Recombinant Antibody [clone: KRTL/7784R]
Catalog # ANTIA316596-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 77
  • Clonality:
    Recombinant
  • Clone:
    KRTL/7784R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q7Z794
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    60 kDa/53 kDa/51 kDa/45 kDa/40 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide corresponding to Type I Cytokeratins residues within aa100-200 of Type I Cytokeratins.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRTL/7784R] antibody to Keratin 77 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Keratin 77
Clonality: Recombinant
Clone: KRTL/7784R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human