Anti-IMP3 Rabbit Recombinant Antibody [clone: IMP3/8934R]
Catalog # ANTIA316567-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BRMS2
- Antigen symbol:IMP3
- Clonality:Recombinant
- Clone:IMP3/8934R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9NV31
- Antigen synonyms:DKFZp686F1078|CT98|DKFZp586L0118|hKOC|FLJ10968|IF2B3_HUMAN|Chromosome 15 open reading frame 12|Cancer/testis antigen 98|C15orf12
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human IMP3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [IMP3/8934R] antibody to IMP3 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: IMP3
Clonality: Recombinant
Clone: IMP3/8934R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human