Order Entry
Netherlands
ContactUsLinkComponent
Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/8287R]
Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/8287R]
Catalog # ANTIA316475-100
Supplier:  Antibodies.com
New Product
Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/8287R]
Catalog # ANTIA316475-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA DP
  • Antigen symbol:
    HLA DR
  • Clonality:
    Recombinant
  • Clone:
    HLA-DRA/8287R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01903
  • Antigen synonyms:
    HLA DR + HLA DP|MHC Class II|HLA DR
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    ~28 kDa (beta chain)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-200) of human HLA-DR protein. The exact sequence is proprietary.
  • Tested applications:
    IHC
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [HLA-DRA/8287R] antibody to HLA DR for IHC with samples derived from human.

  • Validated applications: IHC
  • Recommended dilutions: IHC: (Frozen and Formalin - fixed) 1 to 2 µg/ ml

Type: Primary
Antigen: HLA DR
Clonality: Recombinant
Clone: HLA-DRA/8287R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human