Anti-PGP9.5 Mouse Recombinant Antibody [clone: rUCHL1/8057]
Catalog # ANTIA315511-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Epididymis luminal protein 117
- Antigen symbol:PGP9.5
- Clonality:Recombinant
- Clone:rUCHL1/8057
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P09936
- Antigen synonyms:HEL 117|Neuron cytoplasmic protein 9.5|HEL S 53|Epididymis secretory protein Li 53|OTTHUMP00000218141|OTTHUMP00000218139|OTTHUMP00000218140|NDGOA|OTTHUMP00000218137
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:20 - 30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human UCHL1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rUCHL1/8057] antibody to PGP9.5 for flow cytometry, IF, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: PGP9.5
Clonality: Recombinant
Clone: rUCHL1/8057
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human