Anti-LRP1 Rabbit Recombinant Antibody [clone: LRP1/8947R]
ANTIA315263-100
New Product
- Antibody type:Primary
- Antigen name:A2MR
- Antigen symbol:LRP1
- Clonality:Recombinant
- Clone:LRP1/8947R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q07954
- Antigen synonyms:APOER|APR|Alpha 2 macroglobulin receptor|Apolipoprotein E receptor|alpha 2MR|CD91|CD91 antigen|Alpha-2-macroglobulin receptor|CD 91
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:85 kDa/515 kDa/600 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human LRP1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [LRP1/8947R] antibody to LRP1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: LRP1
Clonality: Recombinant
Clone: LRP1/8947R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human