Bestelling invoeren
Netherlands
ContactUsLinkComponent
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Catalog # ANTIA315213-100
Supplier:  Antibodies.com
New Product
Anti-Keratin 77 Mouse Recombinant Antibody [clone: rKRTL/8751]
Catalog # ANTIA315213-100
Supplier:  Antibodies.com
New Product

Specifications

  • Type antilichaam:
    Primair
  • Clonality:
    Recombinant
  • Clone:
    rKRTL/8751
  • Conjugatie:
    Onconjugeerd
  • Host:
    Mouse
  • ImmunoChemie:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Turtle,
    Rabbit,
    Monkey,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Formulier:
    Liquid
  • Gen-ID:
    UniprotID# Q7Z794
  • Opslagbuffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Moleculair gewicht:
    40 - 56,5 kDa
  • Opslagtemperatuur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentratie:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogeen:
    Human epidermal keratin
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Verpakking:
    Plastic vial
  • VPE:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rKRTL/8751] antibody to Keratin 77 for IHC-P with samples derived from human, monkey, bovine, canine, rabbit, mouse, rat, chicken and turtle.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 0,5 to 1 µg/ml

Type: Primary
Antigen: Keratin 77
Clonality: Recombinant
Clone: rKRTL/8751
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Bovine, Dog, Rabbit, Mouse, Rat, Chicken, Turtle