Order Entry
Netherlands
ContactUsLinkComponent
Anti-Keratin 76 + Keratin 77 Rabbit Recombinant Antibody [clone: 4999R]
Anti-Keratin 76 + Keratin 77 Rabbit Recombinant Antibody [clone: 4999R]
Catalog # ANTIA315211-100
Supplier:  Antibodies.com
New Product
Anti-Keratin 76 + Keratin 77 Rabbit Recombinant Antibody [clone: 4999R]
Catalog # ANTIA315211-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Keratin 76 + Keratin 77
  • Clonality:
    Recombinant
  • Clone:
    4999R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q7Z794 (KRT77), Q01546 (KRT76)
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    40 - 67 kDa (Multiple)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragments and/or synthetic peptides of human Cytokeratin proteins (exact sequences are proprietary)
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit oligoclonal [4999R] antibody to Keratin 76 + Keratin 77 for flow cytometry, IF, WB and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 0,5 to 1 µg/million cells, IF: 1 to 2 µg/ml, WB: 0,5 to 2 µg/ml for 2 hours at RT, IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Keratin 76 + Keratin 77
Clonality: Recombinant
Clone: 4999R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human