Anti-KDM1/LSD1 Mouse Monoclonal Antibody [clone: PCRP-KDM1A-1A10]
ANTIA315209-100
New Product
- Antibody type:Primary
- Antigen name:Amine oxidase (flavin containing) domain 2
- Antigen symbol:KDM1/LSD1
- Clonality:Monoclonal
- Clone:PCRP-KDM1A-1A10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O60341
- Antigen synonyms:EC1|BHC110|BRAF35-HDAC complex protein BHC110|BRAF35 HDAC complex protein BHC110|CPRF|AOF2|Amine oxidase, flavin containing, 2|FAD binding protein BRAF35 HDAC complex, 110 kDa subunit|BRAF35/HDAC complex, 110-kD subunit
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:92,9 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 152-279) of human KDM1A protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-KDM1A-1A10] antibody to KDM1/LSD1 for flow cytometry, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: KDM1/LSD1
Clonality: Monoclonal
Clone: PCRP-KDM1A-1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human