Order Entry
Netherlands
ContactUsLinkComponent
Anti-IMP3 Rabbit Recombinant Antibody [clone: IMP3/8985R]
Anti-IMP3 Rabbit Recombinant Antibody [clone: IMP3/8985R]
  ANTIA315184-100
New Product
Anti-IMP3 Rabbit Recombinant Antibody [clone: IMP3/8985R]
  ANTIA315184-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    BRMS2
  • Antigen symbol:
    IMP3
  • Clonality:
    Recombinant
  • Clone:
    IMP3/8985R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9NV31
  • Antigen synonyms:
    DKFZp686F1078|CT98|DKFZp586L0118|hKOC|FLJ10968|IF2B3_HUMAN|Chromosome 15 open reading frame 12|Cancer/testis antigen 98|C15orf12
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    22 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment of human IMP3 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [IMP3/8985R] antibody to IMP3 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: IMP3
Clonality: Recombinant
Clone: IMP3/8985R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human