Anti-HLA DR Rabbit Recombinant Antibody [clone: HLA-DRA/6840R]
Catalog # ANTIA315093-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:HLA DP
- Antigen symbol:HLA DR
- Clonality:Recombinant
- Clone:HLA-DRA/6840R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01903
- Antigen synonyms:HLA DR + HLA DP|MHC Class II|HLA DR
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:~28 kDa (beta chain)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human HLA-DR protein. The exact sequence is proprietary.
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [HLA-DRA/6840R] antibody to HLA DR for IHC with samples derived from human.
- Validated applications: IHC
- Recommended dilutions: IHC: (Frozen and Formalin to fixed) 1 to 2 µg/ml
Type: Primary
Antigen: HLA DR
Clonality: Recombinant
Clone: HLA-DRA/6840R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human