Order Entry
Netherlands
ContactUsLinkComponent
Anti-CHODL Recombinant Recombinant Antibody [clone: DMC442]
Anti-CHODL Recombinant Recombinant Antibody [clone: DMC442]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-CHODL Recombinant Recombinant Antibody [clone: DMC442]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA318736-500
  • ANTIA318736-100
  • Antigen symbol:
  • CHODL
  • CHODL
  • Antigen name:
  • C21orf68
  • C21orf68
  • Antigen synonyms:
  • CHODL_HUMAN|PRED12|FLJ12627|UNQ872/PRO1890|Chromosome 21 open reading frame 68|CHODL|MT75|Transmembrane protein MT75|Chondrolectin
  • CHODL_HUMAN|PRED12|FLJ12627|UNQ872/PRO1890|Chromosome 21 open reading frame 68|CHODL|MT75|Transmembrane protein MT75|Chondrolectin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DMC442
  • DMC442
  • Reactivity:
  • Human
  • Human
  • Host:
  • Recombinant
  • Recombinant
  • Gene ID:
  • UniprotID# Q9H9P2
  • UniprotID# Q9H9P2
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Form:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Storage temperature:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together