Bestelling invoeren
Netherlands
ContactUsLinkComponent
Anti-GPCR GPR87 Recombinant Recombinant Antibody [clone: DMC478]
Anti-GPCR GPR87 Recombinant Recombinant Antibody [clone: DMC478]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-GPCR GPR87 Recombinant Recombinant Antibody [clone: DMC478]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA318712-500
  • ANTIA318712-100
  • antigeen symbool:
  • GPCR GPR87
  • GPCR GPR87
  • Type antilichaam:
  • Primair
  • Primair
  • Antigeen synoniemen:
  • GPCR87|G-protein coupled receptor 95|G-protein coupled receptor 87|GPCR GPR87|GPR 95|FKSG78|G protein coupled receptor 87|GPR 87|G protein coupled receptor 95
  • GPCR87|G-protein coupled receptor 95|G-protein coupled receptor 87|GPCR GPR87|GPR 95|FKSG78|G protein coupled receptor 87|GPR 87|G protein coupled receptor 95
  • Clonality:
  • Recombinant
  • Recombinant
  • Conjugatie:
  • Onconjugeerd
  • Onconjugeerd
  • Clone:
  • DMC478
  • DMC478
  • Reactivity:
  • Human
  • Human
  • Host:
  • Recombinant
  • Recombinant
  • Gen-ID:
  • UniprotID# Q9BY21
  • UniprotID# Q9BY21
  • Isotype:
  • IgG1
  • IgG1
  • Flowcytometrie:
  • Yes
  • Yes
  • Formulier:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentratie:
  • Reconstitution dependent
  • Reconstitution dependent
  • Opslagbuffer:
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
  • Opslagtemperatuur:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together