Order Entry
Netherlands
ContactUsLinkComponent
Anti-MMP9 Rabbit Recombinant Antibody [clone: MMP9/2025R]
Anti-MMP9 Rabbit Recombinant Antibody [clone: MMP9/2025R]
  ANTIA249377-100
New Product
undefined
Anti-MMP9 Rabbit Recombinant Antibody [clone: MMP9/2025R]
  ANTIA249377-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    82 kDa matrix metalloproteinase-9
  • Antigen symbol:
    MMP9
  • Clonality:
    Recombinant
  • Clone:
    MMP9/2025R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P14780
  • Antigen synonyms:
    92 kDa gelatinase|92 kDa type IV collagenase|Collagenase Type 4 beta|EC 3.4.24.35|Gelatinase 92 KD|CLG 4B|Gelatinase B|Collagenase type IV 92 KD|CLG4B
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    92 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 603-614, of human mMP-9 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [MMP9/2025R] antibody to MMP9 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MMP9
Clonality: Recombinant
Clone: MMP9/2025R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human