Anti-CD99 Mouse Recombinant Antibody [clone: rMIC2/3317]
Catalog # ANTIA249333-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cell surface antigen O13
- Antigen symbol:CD99
- Clonality:Recombinant
- Clone:rMIC2/3317
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|120000000|CD99 molecule|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:27 - 32 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MIC2 protein
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rMIC2/3317] antibody to CD99 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD99
Clonality: Recombinant
Clone: rMIC2/3317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human