Anti-CD99 Mouse Monoclonal Antibody [clone: 12E7 + MIC2/877]
Catalog # ANTIA249332-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:12E7
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:12E7 + MIC2/877
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|CD99 molecule|Cell surface antigen O13|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:27 - 32 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Clone 12E7: Human acute lymphocytic leukemia T-cells. Clone MIC2/877: Recombinant human MIC2 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [12E7 + MIC2/877] antibody to CD99 for flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 5 to 10 µl/million cells, IF: 1:100 to 1:200, IHC-P: 1:100 to 1:200
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: 12E7 + MIC2/877
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human