Anti-EpCAM Mouse Recombinant Antibody [clone: rMOC-31]
Catalog # ANTIA249250-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:17 1A
- Antigen symbol:EpCAM
- Clonality:Recombinant
- Clone:rMOC-31
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P16422
- Antigen synonyms:CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|Cell surface glycoprotein Trop 2|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:40 - 43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Neuraminidase treated GLS-1 human small cell lung carcinoma cells
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rMOC-31] antibody to EpCAM for IF and IHC with samples derived from human.
- Validated applications: IF, IHC
- Recommended dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: EpCAM
Clonality: Recombinant
Clone: rMOC-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human