Anti-HLA DP + DR Mouse Monoclonal Antibody [clone: Bra-14]
Catalog # ANTIA248840-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:HLA DP + DR
- Clonality:Monoclonal
- Clone:Bra-14
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P04440, P01903
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:36 kDa (alpha chain)/27 kDa (beta chain)
- Storage temperature:Shipped at 4 °C. Store at 4 °C.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human REH cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [Bra-14] antibody to HLA DP + DR for flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HLA DP + DR
Clonality: Monoclonal
Clone: Bra-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human