Anti-HIF1 alpha Mouse Monoclonal Antibody [clone: HIF1A/84]
ANTIA248827-100
New Product
- Antibody type:Primary
- Antigen name:ARNT interacting protein
- Antigen symbol:HIF1 alpha
- Clonality:Monoclonal
- Clone:HIF1A/84
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q16665
- Antigen synonyms:HIF 1alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|HIF-1-alpha|HIF-1 alpha|Basic helix loop helix PAS protein MOP1|bHLHe78|HIF 1A|Class E basic helix-loop-helix protein 78
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:92 - 110 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human HIF1 alpha protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [HIF1A/84] antibody to HIF1 alpha for flow cytometry and IF with samples derived from human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: HIF1 alpha
Clonality: Monoclonal
Clone: HIF1A/84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human