Order Entry
Netherlands
ContactUsLinkComponent
Anti-ALK Rabbit Recombinant Antibody [clone: ALK1/2766R]
Anti-ALK Rabbit Recombinant Antibody [clone: ALK1/2766R]
Catalog # ANTIA248626-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-ALK Rabbit Recombinant Antibody [clone: ALK1/2766R]
Catalog # ANTIA248626-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ALK
  • Clonality:
    Recombinant
  • Clone:
    ALK1/2766R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9UM73
  • Antigen synonyms:
    ALK/EML4 fusion gene, included|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK tyrosine kinase receptor|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    80 kDa (hybrid)/200 kDa (wild type)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human ALK protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [ALK1/2766R] antibody to ALK for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: ALK
Clonality: Recombinant
Clone: ALK1/2766R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human