Anti-ALK Mouse Monoclonal Antibody [clone: ALK/1032]
Catalog # ANTIA248623-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:ALK
- Clonality:Monoclonal
- Clone:ALK/1032
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9UM73
- Antigen synonyms:ALK/EML4 fusion gene, included|anaplastic lymphoma kinase (Ki-1)|CD 246|ALK/NPM1 fusion gene, included|Anaplastic lymphoma kinase|anaplastic lymphoma receptor tyrosine kinase|ALK tyrosine kinase receptor|ALK_HUMAN|Anaplastic lymphoma kinase Ki1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:80(hybrid)/200 kDa (wild type)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 220-335, of human ALK protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [ALK/1032] antibody to ALK for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: ALK/1032
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human